Merck
  • Steady-state pharmacokinetics of carvedilol and its enantiomers in patients with congestive heart failure.
Steady-state pharmacokinetics of carvedilol and its enantiomers in patients with congestive heart failure.
Journal of clinical pharmacology (2000-08-10)
D Tenero, S Boike, D Boyle, B Ilson, H F Fesniak, S Brozena, D Jorkasky
ABSTRACT
Carvedilol is a relatively new drug with beta- and alpha 1-receptor blocking activity and antioxidant effects recently approved for the treatment of congestive heart failure (CHF). An ascending, multiple-dose study was completed in 20 male patients with stable New York Heart Association (NYHA) Class III or IV CHF. The pharmacokinetics of carvedilol, S(-)-carvedilol, R(+)-carvedilol, and the active metabolites of carvedilol was assessed at steady state after twice-daily oral administration of carvedilol for 7 days at 6.25, 12.5, 25, and 50 mg doses. Carvedilol exhibited stereoselective pharmacokinetics in CHF patients with dose-proportional increases in steady-state plasma concentrations of carvedilol and its enantiomers. Mean AUC and Cmax values for carvedilol were up to twofold higher in patients with Class IV CHF as compared to those with Class III CHF. Steady-state plasma concentrations of the active metabolites also increased in a dose-proportional manner and were typically 10% or less of that observed for carvedilol. In general, carvedilol was adequately tolerated by adult male CHF patients at the dose levels (6.25-50 mg) evaluated in this study as adverse events were consistent with those frequently observed in patients with CHF.
Sigma-Aldrich
5ā€²-Hydroxy carvedilol, ≥95% (HPLC)
Sigma-Aldrich
4ā€²-Hydroxy carvedilol