Merck
  • Four weeks of exercise early in life reprograms adult skeletal muscle insulin resistance caused by a paternal high-fat diet.
Four weeks of exercise early in life reprograms adult skeletal muscle insulin resistance caused by a paternal high-fat diet.
The Journal of physiology (2018-11-09)
Filippe Falcão-Tebas, Jujiao Kuang, Chelsea Arceri, Jarrod P Kerris, Sofianos Andrikopoulos, Evelyn C Marin, Glenn K McConell
ABSTRACT
A paternal high-fat diet/obesity before mating can negatively influence the metabolism of offspring. Exercise only early in life has a remarkable effect with respect to reprogramming adult rat offspring exposed to detrimental insults before conception. Exercise only early in life normalized adult whole body and muscle insulin resistance as a result of having a high-fat fed/obese father. Unlike the effects on the muscle, early exercise did not normalize the reduced adult pancreatic beta cell mass as a result of having a high-fat fed/obese father. Early-life exercise training may be able to reprogram an individual whose father was obese, inducing long-lasting beneficial effects on health. A paternal high-fat diet (HFD) impairs female rat offspring glucose tolerance, pancreatic morphology and insulin secretion. We examined whether only 4 weeks of exercise early in life could reprogram these negative effects. Male Sprague-Dawley rats consumed a HFD for 10 weeks before mating with chow-fed dams. Female offspring remained sedentary or performed moderate intensity treadmill exercise (5 days week-1 , 60 min day-1 , 20 m min-1 ) from 5 to 9 weeks of age. Paternal HFD impaired (P < 0.05) adult offspring whole body insulin sensitivity (i.p. insulin sensitivity test), as well as skeletal muscle ex vivo insulin sensitivity and TBC1D4 phosphorylation. It also lowered β-cell mass and reduced in vivo insulin secretion in response to an i.p. glucose tolerance test. Early-life exercise in offspring reprogrammed the negative effects of a paternal HFD on whole body insulin sensitivity, skeletal muscle ex vivo insulin-stimulated glucose uptake and TBC1D4 phosphorylation and also increased glucose transporter 4 protein. However, early exercise did not normalize the reduced pancreatic β-cell mass or insulin secretion. In conclusion, only 4 weeks of exercise early in life in female rat offspring reprograms reductions in insulin sensitivity in adulthood caused by a paternal HFD without affecting pancreatic β-cell mass or insulin secretion.
Sigma-Aldrich
Anti-PGC-1α Mouse mAb (4C1.3), liquid, clone 4C1.3, Calbiochem®