Merck
  • Casein kinase 1ε and 1α as novel players in polycystic kidney disease and mechanistic targets for (R)-roscovitine and (S)-CR8.
Casein kinase 1ε and 1α as novel players in polycystic kidney disease and mechanistic targets for (R)-roscovitine and (S)-CR8.
American journal of physiology. Renal physiology (2018-03-15)
Katy Billot, Charlène Coquil, Benoit Villiers, Béatrice Josselin-Foll, Nathalie Desban, Claire Delehouzé, Nassima Oumata, Yannick Le Meur, Alessandra Boletta, Thomas Weimbs, Melanie Grosch, Ralph Witzgall, Sophie Saunier, Evelyne Fischer, Marco Pontoglio, Alain Fautrel, Michal Mrug, Darren Wallace, Pamela V Tran, Marie Trudel, Nikolay Bukanov, Oxana Ibraghimov-Beskrovnaya, Laurent Meijer
ABSTRACT
Following the discovery of (R)-roscovitine's beneficial effects in three polycystic kidney disease (PKD) mouse models, cyclin-dependent kinases (CDKs) inhibitors have been investigated as potential treatments. We have used various affinity chromatography approaches to identify the molecular targets of roscovitine and its more potent analog (S)-CR8 in human and murine polycystic kidneys. These methods revealed casein kinases 1 (CK1) as additional targets of the two drugs. CK1ε expression at the mRNA and protein levels is enhanced in polycystic kidneys of 11 different PKD mouse models as well as in human polycystic kidneys. A shift in the pattern of CK1α isoforms is observed in all PKD mouse models. Furthermore, the catalytic activities of both CK1ε and CK1α are increased in mouse polycystic kidneys. Inhibition of CK1ε and CK1α may thus contribute to the long-lasting attenuating effects of roscovitine and (S)-CR8 on cyst development. CDKs and CK1s may constitute a dual therapeutic target to develop kinase inhibitory PKD drug candidates.
Sigma-Aldrich
Anti-MAP Kinase (ERK-1, 351-368) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
CR8, ≥95% (HPLC)