Merck
  • A signature motif in transcriptional co-activators mediates binding to nuclear receptors.
A signature motif in transcriptional co-activators mediates binding to nuclear receptors.
Nature (1997-06-12)
D M Heery, E Kalkhoven, S Hoare, M G Parker
ABSTRACT
The binding of lipophilic hormones, retinoids and vitamins to members of the nuclear-receptor superfamily modifies the DNA-binding and transcriptional properties of these receptors, resulting in the activation or repression of target genes. Ligand binding induces conformational changes in nuclear receptors and promotes their association with a diverse group of nuclear proteins, including SRC-1/p160, TIF-2/GRIP-1 and CBP/p300 which function as co-activators of transcription, and RIP-140, TIF-1 and TRIP-1/SUG-1 whose functions are unclear. Here we report that a short sequence motif LXXLL (where L is leucine and X is any amino acid) present in RIP-140, SRC-1 and CBP is necessary and sufficient to mediate the binding of these proteins to liganded nuclear receptors. We show that the ability of SRC-1 to bind the oestrogen receptor and enhance its transcriptional activity is dependent upon the integrity of the LXXLL motifs and on key hydrophobic residues in a conserved helix (helix 12) of the oestrogen receptor that are required for its ligand-induced activation function. We propose that the LXXLL motif is a signature sequence that facilitates the interaction of different proteins with nuclear receptors, and is thus a defining feature of a new family of nuclear proteins.
Sigma-Aldrich
SRC1, receptor interaction domain (627-786), GST tagged human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
SRC1, FLAG tagged, TEV site human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
SRC1 (627-786), biotin, untagged human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
SRC1 (627-786), biotin,His tagged human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)