
Verbenalin

Synonym(s):
Verbenaloside
Empirical Formula (Hill Notation):
C17H24O10
CAS Number:
Molecular Weight:
388.37
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing