
Tris(dimethylamino)sulfonium difluorotrimethylsilicate

Synonym(s):
TAS-F, TASF, Tris(dimethylamino)sulfur trimethylsilyl difluoride
Linear Formula:
[(CH3)2N]3S[(CH3)3SiF2]
CAS Number:
Molecular Weight:
275.48
Products
Brand
Formula Weight
Reagent Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing