
Tolterodine L-tartrate

Synonym(s):
2-[(1R)-3-[Bis(1-methylethyl)amino]-1-phenylpropyl]-4-methyl-phenol tartrate, Detrusitol, PNU-200583E
Empirical Formula (Hill Notation):
C22H31NO·C4H6O6
CAS Number:
Molecular Weight:
475.57
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing