![Thymidine powder, BioReagent, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/structures/396/425/8f272551-983b-4797-8cf0-c8c628707feb/640/8f272551-983b-4797-8cf0-c8c628707feb.png)
Thymidine
![Thymidine powder, BioReagent, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/structures/396/425/8f272551-983b-4797-8cf0-c8c628707feb/640/8f272551-983b-4797-8cf0-c8c628707feb.png)
Synonym(s):
1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 2′-Deoxythymidine, Thymine deoxyriboside, dT, Thymidine
Empirical Formula (Hill Notation):
C10H14N2O5
CAS Number:
Molecular Weight:
242.23
Products
Brand
Biological Source
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing