Skip to Content
Merck
Tetraethylammonium borohydride technical, ≥95% (T)

Tetraethylammonium borohydride

Tetraethylammonium borohydride technical, ≥95% (T)
Synonym(s):
NSC 164898, NSC 76086, TEAB, Tetraethylammonium tetrahydroborate
Linear Formula:
(C2H5)4N(BH4)
CAS Number:
Molecular Weight:
145.09
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Reagent Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing