
Sulfamerazine

Synonym(s):
4-Amino-N-(4-methyl-2-pyrimidinyl)benzenesulfonamide, N1-(4-Methylpyrimidin-2-yl)sulfanilamide, Sulfamerazine
Empirical Formula (Hill Notation):
C11H12N4O2S
CAS Number:
Molecular Weight:
264.30
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing