
StratoSpheres™ PL-MPH resin

Synonym(s):
Morpholine, polymer-bound
Empirical Formula (Hill Notation):
C29H33NO
CAS Number:
Molecular Weight:
411.58
Products
Product Category
Brand
Formula Weight
Product Line
Functional Group
Chain Type
Reaction Type
Reactivity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing