
Rosavin

Synonym(s):
(2E)-3-Phenylprop-2-en-1-yl 6-O-α-L-arabinopyranosyl-β-D-glucopyranoside, (2R,3R,4S,5S,6R)-2-[(E)-3-Phenylprop-2-enoxy]-6-([(2S,3R,4S,5S)-3,4,5-trihydroxyoxan-2-yl]oxymethyl)oxane-3,4,5-triol, (E)-3-Phenyl-2-propenyl 6-O-a-L-arabinopyranosyl-β-D-glucopyranoside, Rosavidin
Empirical Formula (Hill Notation):
C20H28O10
CAS Number:
Molecular Weight:
428.43
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing