
Ribavirin impurity standard

Synonym(s):
Ribavirin, 1-β-D-Ribofuranosyl-1,2,4-triazole-3-carboxamide
Empirical Formula (Hill Notation):
C8H12N4O5
CAS Number:
Molecular Weight:
244.20
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing