
Racemic tamsulosin hydrochloride

Synonym(s):
5-[2-[[2-(2-Ethoxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide hydrochloride
Empirical Formula (Hill Notation):
C20H28N2O5S · HCl
CAS Number:
Molecular Weight:
444.97
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing