
Pyrantel Impurity D

Synonym(s):
1,2-Dimethyl-1,4,5,6-tetrahydropyrimidine
Empirical Formula (Hill Notation):
C6H12N2
CAS Number:
Molecular Weight:
112.17
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2