
Punicalin (A + B mixture)

Synonym(s):
Punicalin, mixture of anomers, 4,6-Gallagyl-D-glucose
Empirical Formula (Hill Notation):
C34H22O22
CAS Number:
Molecular Weight:
782.53
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing