Skip to Content
Merck
Potassium hexacyanoferrate(II) trihydrate ACS reagent, 98.5-102.0%

Potassium hexacyanoferrate(II) trihydrate

Potassium hexacyanoferrate(II) trihydrate ACS reagent, 98.5-102.0%
Synonym(s):
Potassium ferrocyanide, Yellow prussiate, Potassium hexacyanoferrate(II) trihydrate
Linear Formula:
K4[Fe(CN)6] · 3H2O
CAS Number:
Molecular Weight:
422.39

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment