Skip to Content
Merck
PMA for use in molecular biology applications, ≥99% (HPLC), Molecular Biology

PMA

PMA for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Synonym(s):
Phorbol 12-myristate 13-acetate, 12-O-Tetradecanoylphorbol 13-acetate, 4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one 12-tetradecanoate 13-acetate, PMA, TPA
Empirical Formula (Hill Notation):
C36H56O8
CAS Number:
Molecular Weight:
616.83

Products

Product Category
Brand
Formula Weight
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing