Skip to Content
Merck
Peptone (vegetable), No. 2 suitable for microbiology

Peptone (vegetable), No. 2

Peptone (vegetable), No. 2 suitable for microbiology
Synonym(s):
Peptone from vegetable
CAS Number:
EC Number:

Products

Product Category
Brand
Biological Source
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing