Partisil™ C18 (5 µm) HPLC Columns
Synonym(s):
Partisil™ HPLC Column
Products
Product Category
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing