
Ofloxacin

Empirical Formula (Hill Notation):
C18H20FN3O4
CAS Number:
Molecular Weight:
361.37
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique