Skip to Content
Merck
Nickel(II) phthalocyanine Dye content 85 %

Nickel(II) phthalocyanine

Nickel(II) phthalocyanine Dye content 85 %
Synonym(s):
Phthalocyanine nickel(II) salt
Empirical Formula (Hill Notation):
C32H16N8Ni
CAS Number:
Molecular Weight:
571.22
EC Number:

Products

Brand
Formula Weight
Chemical Composition
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing