
N-Acetylneuraminic Acid Aldolase from microorganisms

Synonym(s):
N-Acetylneuraminate Pyruvate Lyase, N-Acetylneuraminic Acid Lyase, NANA Aldolase, Sialic Aldolase
CAS Number:
Products
Brand
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing