
Metoprolol Related Compound D

Synonym(s):
(±)-N,N-Bis[2-hydroxy-3-[4-(2-methoxyethyl)phenoxy]propyl](1-methylethyl)amine hydrochloride, 1,1′-[(1-Methylethyl)imino]bis[3-[4-(2-methoxyethyl)phenoxy]-2-propanol hydrochloride
Empirical Formula (Hill Notation):
C27H41NO6 · HCl
CAS Number:
Molecular Weight:
512.08
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2