Merrifield′s peptide resin
Synonym(s):
Chloromethylated polystyrene, Chloromethylated stryene/divinylbenzene copolymer, Chloromethylated styrene-divinylbenzene resin, Copolymer of styrene and divinylbenzene, chloromethylated, Poly(styrene-co-vinylbenzyl chloride-co-divinylbenzene), Polystyrene crosslinked with divinylbenzene, chloromethylated
CAS Number:
Products
Brand
Physical Form
Quality Segment
Application
Cleavage Condition
Linker
Chain Type
Reaction Type
Showing 1-5 of 5
Compare
Product Number
Description
Pricing