
Magnesium carbonate

Synonym(s):
Magnesium carbonate hydrate, Magnesium carbonate
Linear Formula:
MgCO3 · x H2O
Products
Product Category
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique