Skip to Content
Merck
Luperox® A75, Benzoyl peroxide 75%, remainder water

Luperox® A75, Benzoyl peroxide

Luperox® A75, Benzoyl peroxide 75%, remainder water
Synonym(s):
Benzoyl peroxide, Dibenzoyl peroxide
Linear Formula:
(C6H5CO)2O2
CAS Number:
Molecular Weight:
242.23

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing