Skip to Content
Merck
L-Proline SAJ special grade, ≥99.0%

L-Proline

L-Proline SAJ special grade, ≥99.0%
Synonym(s):
(S)-Pyrrolidine-2-carboxylic acid, L-Proline, L-Proline
Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
Amino Acid
API Family
Reference Material Type
Special Grade
Technique