
Lithium hexafluorophosphate

Synonym(s):
Lithium phosphorus fluoride
Linear Formula:
LiPF6
CAS Number:
Molecular Weight:
151.91
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Showing 1-3 of 3