
LiChroprep® Si 60 (15-25 µm)

Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Product Line
Separation Technique
Quality Segment
Application
Particle Size (µm)
Showing 1-1 of 1
Compare
Product Number
Description
Pricing