Skip to Content
Merck
LFM-A13 ≥98% (HPLC), powder

LFM-A13

LFM-A13 ≥98% (HPLC), powder
Synonym(s):
α-Cyano-β-hydroxy-β-methyl-N-(2,5-dibromophenyl)propenamide, (2Z)-2-Cyano-N-(2,5-dibromophenyl)-3-hydroxy-2-butenamide; a-Cyano-b-hydroxy-b-methyl-N-(2,5-dibromophenyl)propenamide
Empirical Formula (Hill Notation):
C11H8Br2N2O2
CAS Number:
Molecular Weight:
360.00

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing