![L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate ≥95%](/deepweb/assets/sigmaaldrich/product/structures/231/444/3dd56c53-f36a-454e-bf11-3ba5cf0b17e5/640/3dd56c53-f36a-454e-bf11-3ba5cf0b17e5.png)
L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate
![L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate ≥95%](/deepweb/assets/sigmaaldrich/product/structures/231/444/3dd56c53-f36a-454e-bf11-3ba5cf0b17e5/640/3dd56c53-f36a-454e-bf11-3ba5cf0b17e5.png)
Empirical Formula (Hill Notation):
C6H6Mg1.5O9P · xH2O
CAS Number:
Molecular Weight:
289.54 (anhydrous basis)
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing