
Lapachol

Synonym(s):
2-Hydroxy-3-(3-methyl-2-butenyl)-1,4-naphthoquinone, Natural Yellow 16
Empirical Formula (Hill Notation):
C15H14O3
CAS Number:
Molecular Weight:
242.27
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing