
Kaempferol 3-glucoside

Synonym(s):
Kaempferol 3-β-D-glucopyranoside, 3,4′,5,7-Tetrahydroxyflavone 3-glucoside, 3-(β-D-Glucopyranosyloxy)-5,7-dihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 3-Glucosylkaempferol, Astragalin, Kaempferol 3-glucoside
Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing