Skip to Content
Merck
Isotonitazene-13C6 hydrochloride solution 100 μg/mL in methanol ((as free base)), certified reference material, ampule of 1 mL, Cerilliant®

Isotonitazene-13C6 hydrochloride solution

Isotonitazene-13C6 hydrochloride solution 100 μg/mL in methanol ((as free base)), certified reference material, ampule of 1 mL, Cerilliant®
Empirical Formula (Hill Notation):
C1713C6H30N4O3 · HCl
Molecular Weight:
452.93

Products

Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Reference Material Type
Special Grade