
Isobutyl methyl ketone

Synonym(s):
4-Methyl-2-pentanone, Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone
Linear Formula:
(CH3)2CHCH2COCH3
CAS Number:
Molecular Weight:
100.16
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Special Grade
Technique
Showing 1-3 of 3