
Ferrous sulfate

Synonym(s):
Iron(II) sulfate heptahydrate, Ferrous sulfate heptahydrate
Linear Formula:
FeSO4 · 7H2O
CAS Number:
Molecular Weight:
278.01
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2