
Etoposide for system suitability

Synonym(s):
Etoposide, 4′-Demethylepipodophyllotoxin 9-(4,6-O-ethylidene-β-D-glucopyranoside), VP-16-213
Empirical Formula (Hill Notation):
C29H32O13
CAS Number:
Molecular Weight:
588.56
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing