
Ethylene glycol dimethacrylate

Synonym(s):
1,2-Ethanediol dimethacrylate, Ethylene dimethacrylate, Ethylene glycol dimethacrylate
Linear Formula:
CH2=C(CH3)COOCH2CH2OCOC(CH3)=CH2
CAS Number:
Molecular Weight:
198.22
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Functionality
Shape
Polymer Composition
Polymer Type
Reaction Type
Reagent Type
Showing 1-2 of 2