
Ethylenediaminetetraacetic acid disodium salt dihydrate

Synonym(s):
Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2, Ethylenediaminetetraacetic acid disodium salt dihydrate
Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
Products
Product Category
Brand
Biological Source
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Research Area
Quality Segment
Application
Reagent Type
Reference Material Type
Special Grade
Technique
Showing 1-13 of 13
Compare
Product Number
Description
Pricing