Skip to Content
Merck
Ethyl acetate JIS 1000, ≥99.5%, for residue analysis

Ethyl acetate

Ethyl acetate JIS 1000, ≥99.5%, for residue analysis
Synonym(s):
EtOAc, Ethyl acetate
Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Special Grade
Technique
Showing 1-20 of 38
Page 1 of 2
Page 1 of 2