Skip to Content
Merck
Ephedrine sulfate United States Pharmacopeia (USP) Reference Standard

Ephedrine sulfate

Ephedrine sulfate United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(−)-Ephedrine hemisulfate salt, (1R,2S)-2-Methylamino-1-phenyl-1-propanol hemisulfate salt, L-(−)-Ephedrine sulfate, L-α-(1-Methylaminoethyl)benzyl alcohol sulfate
Linear Formula:
(C10H15NO)2 · H2SO4
CAS Number:
Molecular Weight:
428.54

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade