Skip to Content
Merck
Docosane certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Docosane

Docosane certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Docosane
Linear Formula:
CH3(CH2)20CH3
CAS Number:
Molecular Weight:
310.60

Products