![DMT-2′O-TC-rU Phosphoramidite configured for PerkinElmer, configured for PolyGen](/deepweb/assets/sigmaaldrich/product/structures/204/830/9e2a29c9-a50b-4973-b8e8-c5fcf2126aa5/640/9e2a29c9-a50b-4973-b8e8-c5fcf2126aa5.png)
DMT-2′O-TC-rU Phosphoramidite
![DMT-2′O-TC-rU Phosphoramidite configured for PerkinElmer, configured for PolyGen](/deepweb/assets/sigmaaldrich/product/structures/204/830/9e2a29c9-a50b-4973-b8e8-c5fcf2126aa5/640/9e2a29c9-a50b-4973-b8e8-c5fcf2126aa5.png)
Synonym(s):
O5′-(4,4′-dimethoxytrityl)-O2′-(1,1-dioxothiomorpholine-4-thiocarbonyl)uridine O3′-(O-(2-cyanoethyl)-N,N-diisopropylphosphoramidite)
Empirical Formula (Hill Notation):
C44H54N5O11PS2
CAS Number:
Molecular Weight:
924.03
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-3 of 3