Skip to Content
Merck
(±)-Propoxyphene-D5 solution 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

(±)-Propoxyphene-D5 solution

(±)-Propoxyphene-D5 solution 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C22D5H24NO2
CAS Number:
Molecular Weight:
344.50

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique