
(±)-Propoxyphene-D11 solution

Synonym(s):
(±)-Propoxyphene-D11 solution
Empirical Formula (Hill Notation):
C22D11H18NO2
Molecular Weight:
350.54
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing