
(±)-Cannabicyclolic Acid (CBLA) solution

Synonym(s):
Cannabicyclolic Acid (CBLA) solution
Empirical Formula (Hill Notation):
C22H30O4
CAS Number:
Molecular Weight:
358.47
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Reference Material Type
Special Grade
Showing 1-1 of 1