![di-Sodium tetraborate decahydrate EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, NF](/deepweb/assets/sigmaaldrich/product/images/311/048/1468862b-2c09-408f-810c-26967205badf/640/1468862b-2c09-408f-810c-26967205badf.jpg)
di-Sodium tetraborate decahydrate
![di-Sodium tetraborate decahydrate EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, NF](/deepweb/assets/sigmaaldrich/product/images/311/048/1468862b-2c09-408f-810c-26967205badf/640/1468862b-2c09-408f-810c-26967205badf.jpg)
Synonym(s):
di-Sodium tetraborate decahydrate
Empirical Formula (Hill Notation):
B4Na2O7 · 10H2O
CAS Number:
Molecular Weight:
381.37
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing