
Dibasic potassium phosphate

Synonym(s):
Potassium phosphate dibasic, Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate
Linear Formula:
K2HPO4
CAS Number:
Molecular Weight:
174.18
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2