
D-Glucose-1,2-13C2

Synonym(s):
Dextrose-1,2-13C2
Empirical Formula (Hill Notation):
13C2C4H12O6
CAS Number:
Molecular Weight:
182.14
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Special Grade
Showing 1-3 of 3